NALG2204-0010 4 Liters Bottle Particulate Cleaned. Materials supplied by: EP. Process: Particulate Cleaning. Packaging: Single cleanroom bag each bottle with cap attached. 6 per case in original box. Certification: C of A USP Particles & Materials Ce 1 * 6 items
Catalog # NALG9FDM4000P-EP
Supplier: Thermo Fisher Scientific
Frequently Bought Together