Temperature control systems, PRESTO®
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen. Multiple interfaces enable remote control of the PRESTO® across networks and in super-ordinated control systems. Wireless remote control of the new PRESTO® can easily be done by using the Wireless TEMP range of products.
- Fingertip control via the touch screen with 14,4 cm colour touch panel
- Up to three user levels with password management
- USB (Host und Device), Ethernet and SD card slot
- Units are whisper-quiet and barely audible when operating
- Filling system is accessible from the top
- Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
A30: 25 L/min, 0,5 bar
A38/A41/W41: 0 to 50 L/min, 0,1 to 1,6 bar
A40/W40, A80/W80: 16 to 40 L/min, 0,3 to 1,7 bar
A45/A45t/W50/W50t: 35 to 76 L/min, 0,48 to 3,2 bar
A70: 0 to 40 L/min, 0,1 to 1,7 bar W91/W92 (t/tt): 26 to 80 L/min, 0,5 to 3,0 bar W91×/W92× (t/tt): 18 to 70 L/min, 0,8 to 5,5 bar
W93: 0 to 80 L/min, 0,1 to 3 bar W93x: 0 to 70 L/min, 0,1 to 5,5 bar.
: Supplied without accessories. Go to website for details of accessories.
: Cooling capacity at +200, +100, +20, 0, –10, –20, –30 and −40 °C. W91/W92/W93 (t/tt/x): Cooling capacity at +200, +100, +20, 0, –20, –40, –60 and −80 °C.
- Heating capacity:15 kW
- Temperature Range:–85...+250 °C
- Weight:335 kg
- Seq:018
- W×D×H:610×845×1250 mm
- Refrigerant:R507
- Cooling capacity:2,8; 2,6; 2,5; 2,4; 2,4; 2,4; 2,2; 0,4 kW
- Temp. stability:±0,05...0,1 °C
- Type:W85t
Accessories
- DescriptionFor
- JULA88900522× adapters, M24×1,5 female to M16×1 maleAll New Presto® series units1 * 2 ST - 153,00 €
- DescriptionFor
- DescriptionFor
- DescriptionFor
- JULA88900672× adapters, M24×1,5 female to M24×1,5 femaleAll New Presto® series units1 * 2 ST - 223,00 €
- DescriptionFor
- JULA88900682× adapters, M24×1,5 female to M30×1,5 maleAll New Presto® series units1 * 2 ST - 246,00 €
- DescriptionFor
- JULA88900692× adapters, M24×1,5 male to M30×1,5 femaleAll New Presto® series units1 * 2 ST - 274,00 €
- DescriptionFor
- JULA88900702× adapters, M24×1,5 female to M30×1,5 femaleAll New Presto® series units1 * 2 ST - 295,00 €
- DescriptionFor
- JULA88900712× adapters, M24×1,5 male to M16×1 femaleAll New Presto® series units1 * 2 ST - 251,00 €
- DescriptionFor
- JULA88900842× adapters, M30×1,5 female to NPT ¾" femaleAll New Presto® series units1 * 2 ST - 215,00 €
- DescriptionFor
- JULA8900020Profibus DP interface, for instruments with RS232All New Presto® series units1 * 1 ST - 1 470,00 €
- DescriptionFor
- JULA8900024RS485 interface, for instruments with RS232All New Presto® series units1 * 1 ST - 1 470,00 €
- DescriptionFor
- JULA8980772Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)All New Presto® series units1 * 1 ST - 576,00 €
- DescriptionFor
- JULA8980773Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)All New Presto® series units1 * 1 ST - 658,00 €
- DescriptionFor
- JULA8900105Electronic module with analog connectors (input, output, standby-in)All New Presto® series units1 * 1 ST - 550,00 €
- DescriptionFor
- JULA8980771Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)All New Presto® series units1 * 1 ST - 555,00 €
- DescriptionFor
- DescriptionFor
- JULA88900802× adapters, M30×1,5 female to M38×1,5 maleAll New Presto® series units1 * 2 ST - 250,00 €
- DescriptionFor
- JULA88900812× adapters, M30×1,5 male to M38×1,5 femaleAll New Presto® series units1 * 2 ST - 250,00 €
- DescriptionFor
- JULA88900832× adapters, M30×1,5 female to NPT ¾" maleAll New Presto® series units1 * 2 ST - 243,00 €
- DescriptionFor
- JULA88900852× adapters, M30×1,5 female to NPT 1" maleAll New Presto® series units1 * 2 ST - 215,00 €
- DescriptionFor
- JULA88900862× adapters, M30×1,5 female to NPT 1" femaleAll New Presto® series units1 * 2 ST - 236,00 €
- DescriptionFor
- DescriptionFor
- DescriptionFor
- JULA88900822× adapters, M30×1,5 female to M38×1,5 femaleAll New Presto® series units1 * 2 ST - 280,00 €
- DescriptionFor
- JULA8970815Sight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 maleAll New Presto® series units1 * 1 ST - 1 450,00 €
- DescriptionFor
- JULA88900622× adapters, M24×1,5 female to NPT 1" maleAll New Presto® series units1 * 2 ST - 214,00 €
- DescriptionFor
- DescriptionFor
- JULA88900612× adapters, M24×1,5 female to NPT ¾" maleAll New Presto® series units1 * 2 ST - 214,00 €
- DescriptionFor
- JULA88900602× adapters, M24×1,5 female to NPT ½" maleAll New Presto® series units1 * 2 ST - 214,00 €
- DescriptionFor
- JULA88900592× adapters, M24×1,5 female to NPT ³/₈" maleAll New Presto® series units1 * 2 ST - 145,00 €
- DescriptionFor
- JULA88900582× adapters, M24×1,5 female to NPT ¼" maleAll New Presto® series units1 * 2 ST - 137,00 €
- DescriptionFor
- JULA88900572× adapters, M24×1,5 female to NPT 1" femaleAll New Presto® series units1 * 2 ST - 274,00 €
- DescriptionFor
- JULA88900562× adapters, M24×1,5 female to NPT ¾" femaleAll New Presto® series units1 * 2 ST - 243,00 €
- DescriptionFor
- JULA88900552× adapters, M24×1,5 female to NPT ½" femaleAll New Presto® series units1 * 2 ST - 217,00 €
- DescriptionFor
- JULA88900542× adapters, M24×1,5 female to NPT ³/₈" femaleAll New Presto® series units1 * 2 ST - 217,00 €
- DescriptionFor
- JULA88900532× adapters, M24×1,5 female to NPT ¼" femaleAll New Presto® series units1 * 2 ST - 153,00 €
- DescriptionFor
- JULA88900892× adapters, M30×1,5 female to M38×1,5 femaleAll New Presto® series units1 * 2 ST - 411,00 €
- DescriptionFor
- JULA88900882× adapters, M30×1,5 female to M30×1,5 femaleAll New Presto® series units1 * 2 ST - 302,00 €
- DescriptionFor
- JULA8900025EtherCAT interface is a protocol converter from EtherCAT to RS232All New Presto® series units1 * 1 ST - 1 470,00 €