1-Propanol ACS
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
- CAS Number:71-23-8
- Formula:H₃CCH₂CH₂OH
- MDL Number:MFCD00002941
- Molecular Weight:60.1 g/mol
- Boiling Pt:97 °C (760 mmHg)
- Melting Pt:–127 °C
- Density:0.8 g/cm³
- Flash Pt:15 °C
- Storage Temperature:Ambient
- UN:1274
- ADR:3,II
Documents
ACRO44577
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.2 %
- Solubility:(in water) Passes test
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Carbonyl compounds:≤0.03 % (as C2H5CHO)
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0004 meq/g
- Ethanol:≤0.01 %
- Methanol:≤0.01 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



