Selenium 99+%, powder
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Selenium 99+%, powder
Catalog # ACRO419275000
Supplier: Thermo Fisher Scientific
CAS Number: 7782-49-2
Jump to:
Product Details & Documents
- CAS Number:7782-49-2
- Formula:Se
- MDL Number:MFCD00134090
- Molecular Weight:78.96 g/mol
- Merck Index:12,08572
- Boiling Pt:∼685 °C (1013 hPa)
- Melting Pt:217 °C
- Density:4.81 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-957-4
- UN:3283
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Dark grey to black Powder
- Assay:≥99.0 %
- Residue after ignition:≤0.5 %
- Copper (Cu):≤100 ppm
- Iron (Fe):≤500 ppm
- Lead (Pb):≤500 ppm
- Nickel (Ni):≤100 ppm
- Sulfur (S):≤500 ppm
Specifications
- Pack type:Glass bottle
- Pk:500 g
- Cat. No.:ACRO419275000
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



