Aluminium acetate basic 90%
Glass bottle, 250 g
Supplier: Thermo Fisher Scientific
Aluminium acetate basic 90%
Catalog # ACRO428192500
Supplier: Thermo Fisher Scientific
CAS Number: 142-03-0
Jump to:
Product Details & Documents
Synonyms: Aluminium Acetate Monobasic
- CAS Number:142-03-0
- Formula:C₄H₇AlO₅
- MDL Number:MFCD00008688
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Aluminium (Al):15.0 to 18.3 % (ICP)
Specifications
- Pack type:Glass bottle
- Pk:250 g



