Order Entry
Austria
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ACS

L(+)-Ascorbic acid ACS

Plastic bottle, 2,5 kg
Catalog # ACRO401470025
L(+)-Ascorbic acid ACS
Catalog # ACRO401470025
CAS Number:  50-81-7
L(+)-Ascorbic acid ACS
L(+)-Ascorbic acid ACS

Plastic bottle, 2,5 kg

CAS Number:  50-81-7
Price
606,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C (dec.)
  • Density:1,694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Iodimetric:
    ≥99.0 %
  • Residue after ignition:
    ≤0.1 %
  • Specific optical rotation:
    +20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (ICP-OES):
    ≤20 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.