Acetylacetone 99+%
Plastic drum, 10 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: ACAC, 2,4-Pentanedione
- CAS Number:123-54-6
- Formula:CH₃COCH₂COCH₃
- MDL Number:MFCD00008787
- Molecular Weight:100.12 g/mol
- Merck Index:13,00083
- Boiling Pt:133…135 °C (760 mmHg)
- Melting Pt:–23 °C
- Density:0.975 g/cm³
- Flash Pt:35.5 °C
- Storage Temperature:Ambient
- EINECS:204-634-0
- UN:2310
- ADR:3,III
Dokumente
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4530 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤50 APHA
- Specific gravity:(20°C) 0.972 to 0.978
- Acidity (CH3COOH):≤0.2 %
Specifications
- Pack type:Plastic drum
- Pk:10 kg



