Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene 99%, extra pure

Mesitylene 99%, extra pure

Glass bottle, 2,5 L
Catalog # ACRO125580025
Irritant
Environment
Flammable
Health
Mesitylene 99%, extra pure
Catalog # ACRO125580025
CAS Number:  108-67-8
Mesitylene 99%, extra pure
Mesitylene 99%, extra pure

Glass bottle, 2,5 L

CAS Number:  108-67-8
Price
248,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:163…166 °C (760 mmHg)
  • Melting Pt:–45 °C
  • Density:0.8675 g/cm³
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4980 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.01 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications