Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Estrone 99+%

Glass bottle, 25 g
Catalog # ACRO117840250
Health
Estrone 99+%
Catalog # ACRO117840250
CAS Number:  53-16-7
Estrone 99+%
Estrone 99+%

Glass bottle, 25 g

CAS Number:  53-16-7
Price
861,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • MDL Number:MFCD00003620
  • Melting Pt:258…261 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-164-5
Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    258 to 261 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 3 hrs)
  • Sulfated ash:
    ≤0.5 %
  • Additional info:
    limit of equilenin and equilin: passes test
  • HPLC:
    ≥99.0 % (on dried substance)
  • Specific optical rotation:
    +158° to +165° (20 °C, 589 nm) (c=1, dioxane)
  • Clarity of solution:
    passes test
  • Silicon dioxide (SiO2):
    ≤0.005 %
  • Sulfur compounds (as SO4):
    ≤0.004 %
  • Insoluble matter:
    ≤0.01 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications