Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol 98+%

D-(-)-Mannitol 98+%

Plastic drum, 5 kg
Catalog # ACRO125340050
D-(-)-Mannitol 98+%
Catalog # ACRO125340050
CAS Number:  69-65-8
D-(-)-Mannitol 98+%
D-(-)-Mannitol 98+%

Plastic drum, 5 kg

CAS Number:  69-65-8
Price
365,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290 - 295 °C (4 hPa)
  • Melting Pt:165…170 °C
  • Density:1,49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    165 to 170 °C
  • Loss on drying:
    ≤0.3 % (105 °C, 4 hrs)
  • HPLC:
    ≥98.0 % (ELSD)
  • Specific optical rotation:
    +23° to +25° (20 °C, 589 nm) (Ph.Eur.-directives)
  • Nickel (Ni):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 kg

Specifications