Microfiber string mops, M-Zone MegaSorb™
MegaSorb™ mop head, 10 oz., irradiated, White, Microcon blue vinyl
Supplier: MICRONOVA
Jump to:
Product Details & Documents
A highly absorbent mop that combines the cleaning efficiency of microfibre with the ease of a looped string mop. The tufted strands are comprised of polyester and nylon filaments, designed to pick up liquids and tiny particles in controlled and critical environments.
- Highly absorbent and low-linting
- Polyester and nylon tufted microfibre tubes
- Picks up tiny particles where cotton fibres just roll over
- White microfibre option includes an additional scrubbing band
- Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
- Compatible with alcohol and strong disinfectants
- Launderable for reuse
- Autoclavable
The M-Zone™ MegaSorb™ Microfibre String Mop features distinctive microfiber tubes with tufted strands made of polyester and nylon filaments, 30 times smaller than cotton fibres, that work to pick up tiny particles.
Specifications
- Colour:White
- Description:MegaSorb™ mop head, 10 oz., irradiated
- Material:Microcon blue vinyl
- Seq:02
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- MicroMop® handle, Fibreglass
Product Information
- Catalog #AvailabilityPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles129-0170Log in for availability1 * 1 ST - 157,00 €
Product Information
- Catalog #LengthMaterialAvailabilityPrice
- Stainless steel quick disconnect extendable handleStainless steel quick disconnect extendable handle129-0551965 - 1829 mmStainless steel 304Log in for availability1 * 1 ST - 358,00 €
Product Information
- Catalog #LengthAvailabilityPrice
- MicroMop® handle, Fibreglass/PP