Orders LinkContactUsLinkComponent
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers with protective film
Catalog # 734-2886
Supplier:   NEXCELOM BIOSCIENCE
Counting chambers, Cellometer®
Catalog # 734-2886
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers with protective film
Price
898,00 €
Log in for availability
1 * 75 ST
Product Details & Documents
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
  • Two independent enclosed chambers
  • Compatible with all Cellometer® cell counters
  • Cell suspension (20 μl) loaded via a a single channel pipette
Specifications
  • Description:
    Counting chambers with protective film
  • Seq:
    01