ContactUsLinkComponent
Multiple sampling adapter with safety cannula AM G 46 K green security Needle 21G x 1 1/2 Suitable for 5ml and 8ml vacuum tubes of different brands. 100 pcs / dispenser box 1000 pcs / carton 1 * 100 ST
  KABE104030
Multiple sampling adapter with safety cannula AM G 46 K green security Needle 21G x 1 1/2 Suitable for 5ml and 8ml vacuum tubes of different brands. 100 pcs / dispenser box 1000 pcs / carton 1 * 100 ST
  KABE104030
Frequently Bought Together