Single Point Controller-X3, CO2-Kohlenstoffdioxid (Infrarot Sensor), 230 VAC, Alarm 1:Relais Output 30 VAC/DC 0,5 A max., Alarm 2:Relais Output 30 VAC/DC 0,5 A max., Alarm 3:interner Warnsummer, Alarm 4:Open Collector Output 30VDC/50mA, Messbereich:0 1 * 1 ST
Catalog # SNSTSPCD31164C100B
Supplier: SENSORTECHNIK TRENKA
Frequently Bought Together