Orders LinkContactUsLinkComponent
Accessories
Accessories
Discharge tube, without recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
Catalog # 613-5207
Supplier:   Brand
Accessories
Catalog # 613-5207
Supplier:  Brand
Accessories
Accessories
Discharge tube, without recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
Price
17,80 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Discharge tube, without recirculation valve, shape: fine tip, 135 mm length, for Dispensette S Organic 25.50 and 100 ml
  • Seq:
    08