Orders LinkContactUsLinkComponent
Accessories
Accessories
Telescoping filling tube, FEP, adjusts to various bottle heights, length 170-330 mm for Dispensette® S/Dispensette® Organic 25,50 and 100 ml
Catalog # 613-5198
Supplier:   Brand
Accessories
Catalog # 613-5198
Supplier:  Brand
Accessories
Accessories
Telescoping filling tube, FEP, adjusts to various bottle heights, length 170-330 mm for Dispensette® S/Dispensette® Organic 25,50 and 100 ml
Price
24,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Telescoping filling tube, FEP, adjusts to various bottle heights, length 170-330 mm for Dispensette® S/Dispensette® Organic 25,50 and 100 ml
  • Seq:
    27