Push button lockout devices
Small (for 16 mm push buttons), Red
118-0527
: Brady
These lockout devices prevent the unauthorised use of push button emergency devices.
- With standard cover
- Made from clear PC, with translucent red cover
- Suitable for 16, 22 or 30 mm push buttons
- Type:Small (for 16 mm push buttons)
- Colour:Red
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



