Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Single channel pipettes, electronic, variable volume, E4™ XLS™

0,5 - 10 µl, 0,01 µl, 2,5 - 1,0 %, 1,2 - 0,4 %, Traditional conical shaft
  RAIN17014491
Marketsource
 :   RAININ
Single channel pipettes, electronic, variable volume, E4™ XLS™
  RAIN17014491
 :  RAININ
Marketsource
Single channel pipettes, electronic, variable volume, E4™ XLS™

0,5 - 10 µl, 0,01 µl, 2,5 - 1,0 %, 1,2 - 0,4 %, Traditional conical shaft

 
2 680,00 €
   
1 * 1 ST
 

The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.

  • Large screen, joystick control and carousel menus speed access to all modes and options
  • True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
  • Independent control of aspirate, dispense and mix speeds for challenging liquids
  • Blowout and automatic dispensing options for precision pipetting protocols
  • Ergonomic fingerhook and joystick provide maximum comfort

Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.

 : Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.

 
  • Volume:
    0,5 - 10 µl
  • With:
    Traditional conical shaft
  • Seq:
    11
  • Increment:
    0,01 µl
  • Accuracy:
    2,5 - 1,0 %
  • Imprecision:
    1,2 - 0,4 %
Accessories

 

  •  
    Description
    For
     
     
  • RAIN30312897
    SmartStand SCS-B
    4× E4™ XLS(+) pipettes
       
    1 * 1 ST - 3 450,00 €