Order Entry
Austria
Orders LinkContactUsLinkComponent
Lead(II) acetate USP Test Solution (TS)

Lead(II) acetate USP Test Solution (TS)

Plastic bottle, 500 mL
Catalog # 85351.260
Supplier:   VWR Chemicals
Lead(II) acetate USP Test Solution (TS)
Catalog # 85351.260
Supplier:  VWR Chemicals
CAS Number:  301-04-2
Lead(II) acetate USP Test Solution (TS)
Lead(II) acetate USP Test Solution (TS)

Plastic bottle, 500 mL

CAS Number:  301-04-2
Price
197,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Ready to use test solution (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Acetic acid Lead(II) salt
  • CAS Number:301-04-2
  • Formula:(H₃CCOO)₂Pb
  • MDL Number:MFCD00012452
  • Molecular Weight:325.29 g/mol
  • Merck Index:12,05411
  • Melting Pt:280 °C
  • EINECS:0
  • UN:1616
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.