Orders LinkContactUsLinkComponent
VWR® Acetate Buffer USP Test Solution (TS)
VWR® Acetate Buffer USP Test Solution (TS)
Acetate buffer TS, Plastic bottle, 500 mL
Catalog # 85323.260
Supplier:   VWR Chemicals
VWR® Acetate Buffer USP Test Solution (TS)
Catalog # 85323.260
Supplier:  VWR Chemicals
CAS Number:  
VWR® Acetate Buffer USP Test Solution (TS)
VWR® Acetate Buffer USP Test Solution (TS)
Acetate buffer TS, Plastic bottle, 500 mL
CAS Number:  
Price
255,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Ready to use test solutions (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Specifications
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Acetate buffer TS