Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution USP Test Solution (TS) 34

Ammonia solution USP Test Solution (TS) 34

Glass bottle, 500 mL
Catalog # 85380.260
Supplier:   VWR Chemicals
Ammonia solution USP Test Solution (TS) 34
Catalog # 85380.260
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution USP Test Solution (TS) 34
Ammonia solution USP Test Solution (TS) 34

Glass bottle, 500 mL

CAS Number:  1336-21-6
Price
103,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Ready to use test solutions (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:0
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications