Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution USP Test Solution (TS) 34

Ammonia solution USP Test Solution (TS) 34

Glass bottle, 100 mL
Catalog # 85380.180
Supplier:   VWR Chemicals
Ammonia solution USP Test Solution (TS) 34
Catalog # 85380.180
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution USP Test Solution (TS) 34
Ammonia solution USP Test Solution (TS) 34

Glass bottle, 100 mL

CAS Number:  1336-21-6
Price
60,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Ready to use test solutions (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:0
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.