Orders LinkContactUsLinkComponent
Samplers, U-D Master
Samplers, U-D Master
Single style sampler, 900 mm, 6, 25 mm
Catalog # SAMP1230A-900
Supplier:   Sampling Systems
Samplers, U-D Master
Catalog # SAMP1230A-900
Supplier:  Sampling Systems
Samplers, U-D Master
Samplers, U-D Master
Single style sampler, 900 mm, 6, 25 mm
Price
5 810,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
U-D Master is designed for sampling free flowing powders. By simply changing the sample insert cells, different volumes can be sampled.
  • Operation of the unit dose powder sampler is very easy
  • Push the sampler: At the required depth rotate the handle, which will take the sample
  • After removing the sampler the sample inserts can be removed for easy emptying of the sample
Single version takes one sample from each location.
Double version takes two samples from each location.
Triple version takes three samples from each location.
Specifications
  • Ø:
    25 mm
  • Description:
    Single style sampler
  • Length:
    900 mm
  • Seq:
    03
  • No. of chambers:
    6
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,2 ml
    Sample insert, stainless steel, 0,2 ml
    SAMP1230T-02
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,9 ml
    Sample insert, stainless steel, 0,9 ml
    SAMP1230T-09
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,0 ml
    Sample insert, stainless steel, 1,0 ml
    SAMP1230T-10
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,1 ml
    Sample insert, stainless steel, 1,1 ml
    SAMP1230T-11
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,3 ml
    Sample insert, stainless steel, 1,3 ml
    SAMP1230T-13
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,2 ml
    Sample insert, stainless steel, 1,2 ml
    SAMP1230T-12
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,4 ml
    Sample insert, stainless steel, 1,4 ml
    SAMP1230T-14
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,5 ml
    Sample insert, stainless steel, 1,5 ml
    SAMP1230T-15
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,6 ml
    Sample insert, stainless steel, 1,6 ml
    SAMP1230T-16
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,7 ml
    Sample insert, stainless steel, 1,7 ml
    SAMP1230T-17
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,8 ml
    Sample insert, stainless steel, 1,8 ml
    SAMP1230T-18
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,9 ml
    Sample insert, stainless steel, 1,9 ml
    SAMP1230T-19
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,0 ml
    Sample insert, stainless steel, 2,0 ml
    SAMP1230T-20
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,2 ml
    Sample insert, stainless steel, 2,2 ml
    SAMP1230T-22
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,3 ml
    Sample insert, stainless steel, 2,3 ml
    SAMP1230T-23
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,4 ml
    Sample insert, stainless steel, 2,4 ml
    SAMP1230T-24
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,5 ml
    Sample insert, stainless steel, 2,5 ml
    SAMP1230T-25
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,6 ml
    Sample insert, stainless steel, 2,6 ml
    SAMP1230T-26
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,7 ml
    Sample insert, stainless steel, 2,7 ml
    SAMP1230T-27
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,9 ml
    Sample insert, stainless steel, 2,9 ml
    SAMP1230T-29
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,0 ml
    Sample insert, stainless steel, 3,0 ml
    SAMP1230T-30
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,2 ml
    Sample insert, stainless steel, 3,2 ml
    SAMP1230T-32
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,3 ml
    Sample insert, stainless steel, 3,3 ml
    SAMP1230T-33
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,1 ml
    Sample insert, stainless steel, 3,1 ml
    SAMP1230T-31
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,4 ml
    Sample insert, stainless steel, 3,4 ml
    SAMP1230T-34
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,5 ml
    Sample insert, stainless steel, 3,5 ml
    SAMP1230T-35
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,6 ml
    Sample insert, stainless steel, 3,6 ml
    SAMP1230T-36
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,8 ml
    Sample insert, stainless steel, 3,8 ml
    SAMP1230T-38
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,9 ml
    Sample insert, stainless steel, 3,9 ml
    SAMP1230T-39
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 4,0 ml
    Sample insert, stainless steel, 4,0 ml
    SAMP1230T-40
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,7 ml
    Sample insert, stainless steel, 3,7 ml
    SAMP1230T-37
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,3 ml
    Sample insert, stainless steel, 0,3 ml
    SAMP1230T-03
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,4 ml
    Sample insert, stainless steel, 0,4 ml
    SAMP1230T-04
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,5 ml
    Sample insert, stainless steel, 0,5 ml
    SAMP1230T-05
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,7 ml
    Sample insert, stainless steel, 0,7 ml
    SAMP1230T-07
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,6 ml
    Sample insert, stainless steel, 0,6 ml
    SAMP1230T-06
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,8 ml
    Sample insert, stainless steel, 2,8 ml
    SAMP1230T-28
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, blank
    Sample insert, stainless steel, blank
    SAMP1230T-00
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,1 ml
    Sample insert, stainless steel, 0,1 ml
    SAMP1230T-01
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,8 ml
    Sample insert, stainless steel, 0,8 ml
    SAMP1230T-08
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,1 ml
    Sample insert, stainless steel, 2,1 ml
    SAMP1230T-21
    Log in for availability
    1 * 1 ST - 152,00 €