Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

4,00, ±0,01, Red, 25 °C, 500 mL
Catalog # REGE1040525C
Supplier:   REAGECON
pH buffer solutions, colour coded
Catalog # REGE1040525C
Supplier:  REAGECON
CAS Number:  
pH buffer solutions, colour coded

4,00, ±0,01, Red, 25 °C, 500 mL

CAS Number:  
Price
32,90 €
Log in for availability
1 * 500 mL
Product Details & Documents

Colour coded pH buffer solutions for the calibration of pH meters.

  • Can be tested at 20 or 25 °C

Certifications: Certified by Reagecon’s ISO 17025 accredited test method and NIST traceable.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • pH accuracy:
    ±0,01
  • pH value:
    4,00

Specifications