Orders LinkContactUsLinkComponent
VWR® Surface Sampling Sponge Kits
VWR® Surface Sampling Sponge Kits
Sterile transport bags and surface sampling sponges with handle pre-moistened with Dey/Engley neutralising buffer
Catalog # 300-0240
Supplier:   VWR Chemicals
VWR® Surface Sampling Sponge Kits
Catalog # 300-0240
Supplier:  VWR Chemicals
VWR® Surface Sampling Sponge Kits
VWR® Surface Sampling Sponge Kits
Sterile transport bags and surface sampling sponges with handle pre-moistened with Dey/Engley neutralising buffer
Price
452,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs
This surface sampling sponge kit is designed to collect and transport samples, to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. These kits are widely used in the food, medical, public health and cosmetic industries.
 : HACCP, USDA, ACIA and CFIA requirements.
 : Gloves should be worn during all sampling procedures.
Specifications
  • Description:
    Sterile transport bags and surface sampling sponges with handle pre-moistened with Dey/Engley neutralising buffer
  • Seq:
    12