Test tubes
10 ml, 95,3 mm, 15,4 mm, graduated, 13,7 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Polypropylene (PP), ultra clear.
- Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
- Centrifugeable up to 15000×g
- Autoclavable
Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.
Specifications
- Ext.Ø:15,4 mm
- Ø int.:13,7 mm
- Version:graduated
- Capacity:10 ml
- Length:95,3 mm
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



