di-Sodium hydrogen phosphate dihydrate EMPROVE® bio Ph Eur,BP,USP This product cannot be returned after delivery 1 * 1 kg
Catalog # 1.37036.1000
Supplier: MERCK PRODUCTION CHEMICALS
di-Sodium hydrogen phosphate dihydrate EMPROVE® bio Ph Eur,BP,USP This product cannot be returned after delivery 1 * 1 kg
Catalog # 1.37036.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
VWR®, Racks, Microcentrifuge Tube
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride low in endotoxins suitable for use as excipient EMPROVE exp Ph Eur,BP,JP,USP 1 * 1 kg
Supplier: MERCK PRODUCTION CHEMICALS
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium dihydrogen phosphate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPE, E339, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals