DNPH reference materials
Glycolaldehyde-2,4-DNPH, 100 µg/ml, CH₃CN, 1 ml/ampoule, 1 Ampoul
Catalog # REST33091
Supplier: Restek
Springe zu:
Product Details & Documents
The reported concentrations reflect the amount of aldehyde or ketone in the mixture.The concentration of derivatised aldehyde or ketone is not reported.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Spezifikationen
Spezifikationen
- Concentration:100 µg/ml
- Matrix:CH₃CN
- Pk:1 Ampoul
- Description:Glycolaldehyde-2,4-DNPH
- Pk Info:1 ml/ampoule



