Orders LinkContactUsLinkComponent
Single - component oxygenates solutions
tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
Catalog # REST30631
Supplier:   Restek
Single - component oxygenates solutions
Catalog # REST30631
Supplier:  Restek
CAS Number:  
Single - component oxygenates solutions
Single - component oxygenates solutions
tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
CAS Number:  
Price
46,00 €
Log in for availability
1 * 1 Ampull
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
  • Pk:
    1 Ampoul
  • Standards:
    US EPA 502.2, 8021, 8206B
  • Description:
    tert-Amyl alcohol, with 10000 µg/ml in purge and trap grade methanol
  • Pk Info:
    1 ml/ampoule
Specifications