Single - component oxygenates solutions
Ethyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
Catalog # REST30628
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pk:1 Ampoul
- Standards:US EPA 502.2, 8021, 8206B
- Description:Ethyl tert-butyl ether (ETBE), with 2000 µg/ml in purge and trap grade methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30628
- Formula:C₆H₁₄O
- Description:Ethyl-tert-butyl ether (ETBE)
- CAS No.:637-92-3



