Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Resprep®, method specific
Solid phase extraction cartridges, Resprep®, method specific
Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquat, PP, 6 ml, 500 mg
Catalog # REST25499
Supplier:   Restek
Solid phase extraction cartridges, Resprep®, method specific
Catalog # REST25499
Supplier:  Restek
Solid phase extraction cartridges, Resprep®, method specific
Solid phase extraction cartridges, Resprep®, method specific
Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquat, PP, 6 ml, 500 mg
Price
289,00 €
Log in for availability
1 * 30 ST
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifications
  • Capacity:
    6 ml
  • Description:
    Ultra Quat SPE method, for use in HPLC analysis of paraquat/diquat
  • Seq:
    01
  • Bed weight:
    500 mg
  • Column hardware:
    PP