Order Entry
Austria
Orders LinkContactUsLinkComponent
Conductivity cells, IDS

Conductivity cells, IDS

IDS conductivity cell, TetraCon® 925/C, 4-cell graphite, PEEK head, TetraCon® 925/C, 4-cell graphite, PEEK head, 1 µS/cm - 2000 mS/cm, 0,475 cm-1, 0...+100 °C, PEEK, 15,3×120 mm, IDS-plug, 1,5 m cable
Catalog # 665-0475
Supplier:   WTW
Conductivity cells, IDS
Catalog # 665-0475
Supplier:  WTW
Conductivity cells, IDS
Conductivity cells, IDS

IDS conductivity cell, TetraCon® 925/C, 4-cell graphite, PEEK head, TetraCon® 925/C, 4-cell graphite, PEEK head, 1 µS/cm - 2000 mS/cm, 0,475 cm-1, 0...+100 °C, PEEK, 15,3×120 mm, IDS-plug, 1,5 m cable

Price
991,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.

  • Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • IDS cells store their serial number and calibration records
  • TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
  • LR 925/01: Ideal for pure water and low conductivity applications
  • TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples

The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.

Ordering information: Various cable lengths available.

Specifications
  • Conductivity:
    1 µS/cm - 2000 mS/cm
  • Connection:
    IDS-plug, 1,5 m cable
  • Ø×L:
    15,3×120 mm
  • Type:
    TetraCon® 925/C, 4-cell graphite, PEEK head
  • Description:
    IDS conductivity cell, TetraCon® 925/C, 4-cell graphite, PEEK head
  • Material:
    PEEK
  • Seq:
    07
  • Temp.:
    0...+100 °C
  • Cell constant:
    0,475 cm-1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.