Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/17, 0,25 ± ml
Catalog # POULRB258/WAC/J/2
Supplier:   Poulten & Graf
Volumetric flasks, class A
Catalog # POULRB258/WAC/J/2
Supplier:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/17, 0,25 ± ml
Price
51,30 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifications
  • NS:
    19/17
  • Capacity:
    500 ml
  • Seq:
    14
  • Tolerance:
    0,25 ± ml