Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
50 ml, 12/14, 0,06 ± ml
Catalog # POULRB258/WAC/F/5
Supplier:   Poulten & Graf
Volumetric flasks, class A
Catalog # POULRB258/WAC/F/5
Supplier:  Poulten & Graf
Volumetric flasks, class A
Volumetric flasks, class A
50 ml, 12/14, 0,06 ± ml
Price
73,80 €
Log in for availability
1 * 5 ST
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
Specifications
  • NS:
    12/14
  • Capacity:
    50 ml
  • Seq:
    07
  • Tolerance:
    0,06 ± ml