Volumetric flasks, class A
1000 ml, 24/20, 0,40 ± ml
Catalog # POULRB258/K/2
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Specifications
- NS:24/20
- Capacity:1000 ml
- Seq:28
- Tolerance:0,40 ± ml