E-Z 96® Lysate Clearance Plate 1 * 10 items
Catalog # FL9601
Supplier: OMEGA BIO-TEK
Frequently Bought Together
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, dNTPs
Supplier: VWR Chemicals
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
DNA ladders, GeneRuler™ and O'GeneRuler™
Supplier: Thermo Fisher Scientific
High Sensitivity DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
NUCLEOSPIN BUFFER A2 1 L 1 * 1 KIT
Supplier: MACHEREY-NAGEL
VWR®, Hot Start PCR-to-Gel Taq PCR master mix, 2X
Supplier: VWR Chemicals
DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Individual DNA fragment NoLimits 700bp DNA Fragment 10µg (0,5µg/µl) 1 * 1 KIT
Supplier: Thermo Fisher Scientific
DNA loading dye TriTrack 6x 5x1ml 1 * 1 KIT
Supplier: Thermo Fisher Scientific
Buffer CF (1000 mL) Lysis Buffer CF bottle of 1000 mL 1 * 1.000 mL
Supplier: MACHEREY-NAGEL
Column end plugs
Supplier: VICI JOUR RESEARCH
RNA ScreenTape Analysis
Supplier: AGILENT