CELLOMETER AUTO M10 Automatic Cell Counter incl. PC Microscope-based automated cell counter with CCD camera. The system determines cell concentration and cell size in only 20 µl growth medium or buffer without staining. Viability assays after Trypan 1 * 1 items
Catalog # NEXC91-CMM10-C
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together