Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample Bags without Closure

Sample Bags without Closure

1340×800 mm, 0,08 mm
Catalog # SCAC2080
Sample Bags without Closure
Catalog # SCAC2080
Sample Bags without Closure
Sample Bags without Closure

1340×800 mm, 0,08 mm

Price
231,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Bags made from PE transparent, for the storage or transport of samples.

Available in several sizes.

Caution: Supplied without closure ties.

Specifications
  • L×W:
    1340×800 mm
  • Seq:
    22
  • Thickness:
    0,08 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.