ContactUsLinkComponent
HWS SEAL RING STIRRING CLOSURE, NEW, GUIDE SLEEVE AND BODY MADE OF PEEK SEALS MADE OF VITON FOR SHAFT Ø 10 MM, GLASS ADAPTER DN 25 P 1 * 1 items
Catalog # HWSA3792325
Supplier:  HWS LABORTECHNIK
HWS SEAL RING STIRRING CLOSURE, NEW, GUIDE SLEEVE AND BODY MADE OF PEEK SEALS MADE OF VITON FOR SHAFT Ø 10 MM, GLASS ADAPTER DN 25 P 1 * 1 items
Catalog # HWSA3792325
Supplier:  HWS LABORTECHNIK
Frequently Bought Together