ContactUsLinkComponent
Hydrometer precision for extract and wine, range 0.985-1.003g/ml, div 0.0002, length 335mm, accuracy +/-0.0004 calibraion temp 20°C. 1 * 1 items
Catalog # ALLA2960FC098/20QP
Supplier:  Alla France
Hydrometer precision for extract and wine, range 0.985-1.003g/ml, div 0.0002, length 335mm, accuracy +/-0.0004 calibraion temp 20°C. 1 * 1 items
Catalog # ALLA2960FC098/20QP
Supplier:  Alla France
Frequently Bought Together