ContactUsLinkComponent
The CMC-535 Detergent Assay is a simple, fluorescent assay designed for the detection of various detergents in aqueous solutions a nd is an ideal compliment for the DetergentOUT detergent removal columns. The basic principle of the assay is the in 1 * 1 KIT
Catalog # GENODG535
Supplier:  G-Biosciences
The CMC-535 Detergent Assay is a simple, fluorescent assay designed for the detection of various detergents in aqueous solutions a nd is an ideal compliment for the DetergentOUT detergent removal columns. The basic principle of the assay is the in 1 * 1 KIT
Catalog # GENODG535
Supplier:  G-Biosciences