Mode of action and Anti-microbial spectrum: Cefotaxime is an inhibitor of bacterial cell wall synthesis. High activity against gram- negative bacteria. Very often used for the elimination of Agrobacterium species after inoculation. High resistance ag 1 * 1 g
Catalog # GENORC-164
Supplier: G-Biosciences
Frequently Bought Together