CYPROHEPTADINE HYDROCHLORIDE 1 * 30 mg
Catalog # EDQMC3280000
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Cinnamaldehyde, Reference Standard
Supplier: USP
Viscosity standards, general purpose
Supplier: CANNON INSTRUMENT
Quinine Sulfate, Reference Standard
Supplier: USP
7H-BENZO(C)FLUORENE 1 * 10 mg
Supplier: EHRENSTORFER
Organic reference standard, N-nitroso-di-isopropylamine, Dr Ehrenstorfer
Supplier: EHRENSTORFER
pH buffer solutions, colour coded
Supplier: Hach
2-Naphthylamine, Supelco®
Supplier: Merck
Accessories for conductivity meters, Portavo
Supplier: KNICK
Metaraminol Related Compounds, Reference Standard
Supplier: USP
Supelco® PESTANAL® Organic Reference Standard, Tebuconazole
Supplier: Merck
Supelco® Organic Reference Standard, 3-Methylbutanol
Supplier: Merck
FLUFENZINE,ANALYTICAL STANDARD 1 * 0,1 g
Supplier: EHRENSTORFER
SILDENAFIL 1 * 10 mg
Supplier: LGC Standards PROMOCHEM
Furosemide, Reference Standards
Supplier: USP