Order Entry
Austria
Orders LinkContactUsLinkComponent
Swing samplers

Swing samplers

Swing sampler, up to 365 cm
Catalog # NASCB01310WA
Supplier:   NASCO FORT ATKINSON
Swing samplers
Catalog # NASCB01310WA
Swing samplers
Swing samplers

Swing sampler, up to 365 cm

Price
623,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

For collecting liquids, powders, or small solids from a horizontal flowing stream

The end of the sampler swings to allow collection from different angles, including 90°. Samplers come with an extending fiberglass pole and one 960mL (32.5oz.) polyethylene bottle with plastic snapper ring.

Specifications
  • Description:
    Swing sampler, up to 365 cm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.