BUFFER SOLUTION PH9.18/ 0.01/ 25C 1 * 500 mL
Catalog # REGEEP918
Supplier: REAGECON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Pipette Tips, Traditional
Supplier: RAININ