Conductivity standards
Conductivity standard solution, 1413 µS/cm, 250 mL
Supplier: Hach
Jump to:
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifications
Specifications
- Pk:250 mL
- Description:Conductivity standard solution
- Conductivity:1413 µS/cm