FLUCONAZOLE IMPURITY C 5MG 1 * 2 items
Catalog # EDQMY0000574
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions
Supplier: Hanna
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer, single-use, SINGLET™
Supplier: Hach