Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Sampling spoons, LaboPlast

127 mm, 2,5 ml
  231-0513
 :   BURKLE
Sampling spoons, LaboPlast
  231-0513
 :  BURKLE
Sampling spoons, LaboPlast
Sampling spoons, LaboPlast

127 mm, 2,5 ml

 
42,50 €
   
1 * 100 ST
 

PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.

  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed
 
  • Capacity:
    2,5 ml
  • Length:
    127 mm
  • Seq:
    01