Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, Sampling Spoons

VWR®, Sampling Spoons

127 mm, 2,5 ml
Catalog # 231-0493
Supplier:   VWR Collection
VWR®, Sampling Spoons
Catalog # 231-0493
Supplier:  VWR Collection
VWR®, Sampling Spoons
VWR®, Sampling Spoons

127 mm, 2,5 ml

Price
49,60 €
Log in for availability
1 * 100 ST
Product Details & Documents

PS, white. These disposable spoons are produced in a cleanroom environment and sterilised by gamma radiation. They are ideal for sampling powder, granules and pastes.

  • Available in two different sizes: 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon)
  • Production, assembly and packaging according to cleanroom Class 7 (10000)
  • According to EU food and FDA guidelines
  • Individually packed, sterilised by gamma radiation
Specifications
  • Capacity:
    2,5 ml
  • Length:
    127 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.