VWR®, Sampling Scoops
150 ml, 216 mm, White
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, white. These sterile scoops are ideal for sampling powder, granules and pastes.
- Individually packed
- Sterilised by gamma radiation
Thanks to the special handle construction, and the slightly angled edge, the scoop stands horizontally and can be laid down without the material trickling out.
: Production, assembly and packaging according to cleanroom Class 7 (10000). According to EU food and FDA guidelines.
: *The product(s) below are outside the scope of the EU Directive 2019/904.
Documents
Specifications
- Capacity:150 ml
- Colour:White
- Length:216 mm
- Seq:05



